3-Fluoro-5-(trifluoromethoxy)-DL-phenylalanine
Supplier: Apollo Scientific
PC302848-1G PC302848-5G
APOSPC302848-1GEA 238 EUR
APOSPC302848-1G APOSPC302848-5G
3-Fluoro-5-(trifluoromethoxy)-DL-phenylalanine
3-Fluoro-5-(trifluoromethoxy)-DL-phenylalanine
Formula: C₁₀H₉F₄NO₃
MW: 267.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783233
CAS Number: 1391002-18-8