2-Methoxy-4,6-bis(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
PC302926-10G PC302926-5G PC302926-1G
APOSPC302926-10GEA 0 EUR
APOSPC302926-10G APOSPC302926-5G APOSPC302926-1G
2-Methoxy-4,6-bis(trifluoromethyl)benzoyl chloride 97%
2-Methoxy-4,6-bis(trifluoromethyl)benzoyl chloride
Formula: C₁₀H₅ClF₆O₂
MW: 306.589 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00729119
CAS Number: 886503-47-5