3-(2,3,5-Trifluorophenyl)propionic acid 98%
Supplier: Apollo Scientific
PC302936-10G PC302936-5G PC302936-1G
APOSPC302936-10GEA 316 EUR
APOSPC302936-10G APOSPC302936-5G APOSPC302936-1G
3-(2,3,5-Trifluorophenyl)propionic acid 98%
3-(2,3,5-Trifluorophenyl)propionic acid
Formula: C₉H₇F₃O₂
MW: 204.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660215
CAS Number: 886499-45-2