3-Butoxy-2,6-difluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC302942-10G PC302942-5G PC302942-1G
APOSPC302942-10GEA 316 EUR
APOSPC302942-10G APOSPC302942-5G APOSPC302942-1G
3-Butoxy-2,6-difluorobenzoic acid 97%
3-Butoxy-2,6-difluorobenzoic acid
Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986833
CAS Number: 1706434-98-1