3-Butoxy-2,6-difluorophenylacetonitrile 97%
Supplier: Apollo Scientific
Danger
PC302943-1G PC302943-5G
APOSPC302943-1GEA 180 EUR
APOSPC302943-1G APOSPC302943-5G
3-Butoxy-2,6-difluorophenylacetonitrile 97%
3-Butoxy-2,6-difluorophenylacetonitrile
Formula: C₁₂H₁₃F₂NO
MW: 225.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986843
CAS Number: 1706435-04-2
UN: 3439
ADR: 6.1,III