4-Chloro-3-(trifluoromethoxy)benzoyl chloride, tech
Supplier: Apollo Scientific
PC302977-10G PC302977-5G PC302977-1G
APOSPC302977-10GEA 405 EUR
APOSPC302977-10G APOSPC302977-5G APOSPC302977-1G
4-Chloro-3-(trifluoromethoxy)benzoyl chloride, tech
4-Chloro-3-(trifluoromethoxy)benzoyl chloride
Formula: C₈H₃Cl₂F₃O₂
MW: 259.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660239
CAS Number: 886500-55-6