5-Methyl-2-(trifluoromethoxy)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
PC303072-1G PC303072-250MG
APOSPC303072-1GEA 2210 EUR
APOSPC303072-1G APOSPC303072-250MG
5-Methyl-2-(trifluoromethoxy)benzenesulphonyl chloride 97%
5-Methyl-2-(trifluoromethoxy)benzenesulphonyl chloride
Formula: C₈H₆ClF₃O₃S
MW: 274.645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399740
CAS Number: 1261683-44-6