3,6-Dichloro-2-fluoro-DL-phenylalanine
Supplier: Apollo Scientific
PC303122-1G
PC303122-10G
PC303122-5G
APOSPC303122-1GEA
259
EUR
APOSPC303122-1G
APOSPC303122-10G
APOSPC303122-5G
3,6-Dichloro-2-fluoro-DL-phenylalanine
3,6-Dichloro-2-fluoro-DL-phenylalanine
Formula:
C₉H₈Cl₂FNO₂ MW: 252.07 g/mol Melting Pt: 265 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832308 CAS Number: 1256482-67-3 |