2,3-Difluoro-4-pentylphenol 98%
Supplier: Apollo Scientific
PC303131-1G PC303131-10G PC303131-5G
APOSPC303131-1GEA 296 EUR
APOSPC303131-1G APOSPC303131-10G APOSPC303131-5G
2,3-Difluoro-4-pentylphenol 98%
2,3-Difluoro-4-pentylphenol
Formula: C₁₁H₁₄F₂O
Storage Temperature: Ambient
MDL Number: MFCD09258677
CAS Number: 887582-85-6
UN: 1759
ADR: 8,III