3,4-Dichloro-5-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC303159-1G PC303159-250MG
APOSPC303159-1GEA 528 EUR
APOSPC303159-1G APOSPC303159-250MG
3,4-Dichloro-5-(trifluoromethyl)benzaldehyde 98%
3,4-Dichloro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054198
CAS Number: 1686130-35-7