2-Chloro-5-fluoro-3-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
PC303163-1G PC303163-250MG
APOSPC303163-1GEA 574 EUR
APOSPC303163-1G APOSPC303163-250MG
2-Chloro-5-fluoro-3-(trifluoromethyl)aniline 98%
2-Chloro-5-fluoro-3-(trifluoromethyl)aniline
Formula: C₇H₄ClF₄N
MW: 213.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986847
CAS Number: 105172-78-9
UN: 2810
ADR: 6.1,III