2-Methyl-3-(trifluoromethyl)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Methyl-3-(trifluoromethyl)benzene-1-sulphonyl chloride
PC303182-1G PC303182-5G
APOSPC303182-1GEA 398 EUR
APOSPC303182-1G APOSPC303182-5G
2-Methyl-3-(trifluoromethyl)benzenesulphonyl chloride 97%
2-Methyl-3-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₈H₆ClF₃O₂S
MW: 258.645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026169
CAS Number: 1191545-46-6
UN: 3265
ADR: 8,II