3,6-Dichloro-2-fluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
PC303184-1G PC303184-250MG
APOSPC303184-1GEA 676 EUR
APOSPC303184-1G APOSPC303184-250MG
3,6-Dichloro-2-fluorobenzenesulphonyl chloride 97%
3,6-Dichloro-2-fluorobenzenesulphonyl chloride
Formula: C₆H₂Cl₃FO₂S
MW: 263.501 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054224
CAS Number: 1706436-29-4
UN: 1760
ADR: 8,III