2,3-Difluoro-5-methoxy-4-methylbenzoic acid 97%
Supplier: Apollo Scientific
PC303200-1G PC303200-250MG
APOSPC303200-1GEA 788 EUR
APOSPC303200-1G APOSPC303200-250MG
2,3-Difluoro-5-methoxy-4-methylbenzoic acid 97%
2,3-Difluoro-5-methoxy-4-methylbenzoic acid
Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986839
CAS Number: 1706446-21-0