3,4-Dichloro-5-(trifluoromethoxy)aniline 97%
Supplier: Apollo Scientific
PC303205-1G PC303205-250MG
APOSPC303205-1GEA 788 EUR
APOSPC303205-1G APOSPC303205-250MG
3,4-Dichloro-5-(trifluoromethoxy)aniline 97%
3,4-Dichloro-5-(trifluoromethoxy)aniline
Formula: C₇H₄Cl₂F₃NO
MW: 246.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054197
CAS Number: 1706430-28-5