2,3-Difluoro-6-methoxybenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
PC303226-1G PC303226-250MG
APOSPC303226-1GEA 899 EUR
APOSPC303226-1G APOSPC303226-250MG
2,3-Difluoro-6-methoxybenzenesulphonyl chloride 97%
2,3-Difluoro-6-methoxybenzenesulphonyl chloride
Formula: C₇H₅ClF₂O₃S
MW: 242.628 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922651
CAS Number: 1208074-96-7