3,4-Dichloro-5-(trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
PC303232-1G PC303232-250MG
APOSPC303232-1GEA 899 EUR
APOSPC303232-1G APOSPC303232-250MG
3,4-Dichloro-5-(trifluoromethyl)phenylacetonitrile 97%
3,4-Dichloro-5-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₄Cl₂F₃N
MW: 254.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103372
CAS Number: 1824274-76-1
UN: 3439
ADR: 6.1,III