4-Fluoro-3,5-dimethylbenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
PC303235-1G PC303235-250MG
APOSPC303235-1GEA 619 EUR
APOSPC303235-1G APOSPC303235-250MG
4-Fluoro-3,5-dimethylbenzenesulphonyl chloride 98%
4-Fluoro-3,5-dimethylbenzenesulphonyl chloride
Formula: C₈H₈ClFO₂S
MW: 222.664 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19201132
CAS Number: 122263-78-9
UN: 1760
ADR: 8 III