2,3-Difluoro-5-methoxy-4-methylbenzyl bromide 97%
Supplier: Apollo Scientific
PC303243-1G PC303243-250MG
APOSPC303243-1GEA 1020 EUR
APOSPC303243-1G APOSPC303243-250MG
2,3-Difluoro-5-methoxy-4-methylbenzyl bromide 97%
2,3-Difluoro-5-methoxy-4-methylbenzyl bromide
Formula: C₉H₉BrF₂O
MW: 251.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986851
CAS Number: 1706430-78-5
UN: 3265
ADR: 8 III