2-Chloro-6-(trifluoromethyl)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
PC303244-1G PC303244-5G
APOSPC303244-1GEA 270 EUR
APOSPC303244-1G APOSPC303244-5G
2-Chloro-6-(trifluoromethyl)benzenesulphonyl chloride 97%
2-Chloro-6-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃Cl₂F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD06408806
CAS Number: 874814-70-7
UN: 1759
ADR: 8,II