5-Ethoxy-2,3-difluoro-4-methylbenzaldehyde 97%
Supplier: Apollo Scientific
PC303253-1G PC303253-250MG
APOSPC303253-1GEA 1040 EUR
APOSPC303253-1G APOSPC303253-250MG
5-Ethoxy-2,3-difluoro-4-methylbenzaldehyde 97%
5-Ethoxy-2,3-difluoro-4-methylbenzaldehyde
Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986848
CAS Number: 1706446-37-8