2,3-Difluoro-5-methoxy-4-methylphenylacetic acid 98%
Supplier: Apollo Scientific
PC303256-1G PC303256-250MG
APOSPC303256-1GEA 1160 EUR
APOSPC303256-1G APOSPC303256-250MG
2,3-Difluoro-5-methoxy-4-methylphenylacetic acid 98%
2,3-Difluoro-5-methoxy-4-methylphenylacetic acid
Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986854
CAS Number: 1706430-83-2