2-Ethoxy-3,5-difluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
PC303259-1G PC303259-250MG
APOSPC303259-1GEA 1230 EUR
APOSPC303259-1G APOSPC303259-250MG
2-Ethoxy-3,5-difluorobenzenesulphonyl chloride 97%
2-Ethoxy-3,5-difluorobenzenesulphonyl chloride
Formula: C₈H₇ClF₂O₃S
MW: 256.654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19434751
CAS Number: 1487710-75-7
UN: 1760
ADR: 8,III