2,4-Difluoro-5-methylbenzamide 97%
Supplier: Apollo Scientific
PC303308-5G PC303308-1G
APOSPC303308-5GEA 874 EUR
APOSPC303308-5G APOSPC303308-1G
2,4-Difluoro-5-methylbenzamide 97%
2,4-Difluoro-5-methylbenzamide
Formula: C₈H₇F₂NO
MW: 171.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19440203
CAS Number: 1379078-33-7