2,4-Difluoro-5-methylbenzenesulphonamide 97%
Supplier: Apollo Scientific
PC303310-1G PC303310-250MG
APOSPC303310-1GEA 493 EUR
APOSPC303310-1G APOSPC303310-250MG
2,4-Difluoro-5-methylbenzenesulphonamide 97%
2,4-Difluoro-5-methylbenzenesulphonamide
Formula: C₇H₇F₂NO₂S
MW: 207.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22369817
CAS Number: 1384430-88-9