2,4-Difluoro-5-methylbenzylamine 98%
Supplier: Apollo Scientific
PC303311-1G PC303311-5G
APOSPC303311-1GEA 377 EUR
APOSPC303311-1G APOSPC303311-5G
2,4-Difluoro-5-methylbenzylamine 98%
2,4-Difluoro-5-methylbenzylamine
Formula: C₈H₉F₂N
MW: 157.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18825364
CAS Number: 329314-63-8