2,5-Dichloro-4-fluoroaniline 98%
Supplier: Apollo Scientific
PC303313-5G PC303313-1G PC303313-10G
APOSPC303313-5GEA 120 EUR
APOSPC303313-5G APOSPC303313-1G APOSPC303313-10G
2,5-Dichloro-4-fluoroaniline 98%
2,5-Dichloro-4-fluoroaniline
Formula: C₆H₄Cl₂FN
MW: 180.007 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06808808
CAS Number: 2729-37-5
UN: 2811
ADR: 6.1,III