3,6-Difluoro-2-methoxybenzamide 97%
Supplier: Apollo Scientific
PC303319-5G PC303319-1G
APOSPC303319-5GEA 1300 EUR
APOSPC303319-5G APOSPC303319-1G
3,6-Difluoro-2-methoxybenzamide 97%
3,6-Difluoro-2-methoxybenzamide
Formula: C₈H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD18394893
CAS Number: 1261439-98-8