4-Chloro-2-fluoro-5-methylbenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
PC303320-5G
PC303320-1G
APOSPC303320-5GEA
717
EUR
APOSPC303320-5G
APOSPC303320-1G
4-Chloro-2-fluoro-5-methylbenzaldehyde
4-Chloro-2-fluoro-5-methylbenzaldehyde
Formula:
C₈H₆ClFO MW: 172.584 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28751755 CAS Number: 177211-30-2 |