2-Chloro-4-fluoro-5-methylbenzonitrile 97%
Supplier: Apollo Scientific
PC303345-1G PC303345-5G
APOSPC303345-1GEA 273 EUR
APOSPC303345-1G APOSPC303345-5G
2-Chloro-4-fluoro-5-methylbenzonitrile 97%
2-Chloro-4-fluoro-5-methylbenzonitrile
Formula: C₈H₅ClFN
MW: 169.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21602109
CAS Number: 924626-79-9