2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
PC303382-1G PC303382-250MG
APOSPC303382-1GEA 592 EUR
APOSPC303382-1G APOSPC303382-250MG
2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile 97%
2-Fluoro-3-methyl-5-(trifluoromethyl)benzonitrile
Formula: C₉H₅F₄N
MW: 203.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31916395
CAS Number: 2383627-27-6