2-Fluoro-3-methyl-5-(trifluoromethyl)phenol 98%
Supplier: Apollo Scientific
PC303385-1G PC303385-250MG
APOSPC303385-1GEA 681 EUR
APOSPC303385-1G APOSPC303385-250MG
2-Fluoro-3-methyl-5-(trifluoromethyl)phenol 98%
2-Fluoro-3-methyl-5-(trifluoromethyl)phenol
Formula: C₈H₆F₄O
MW: 194.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640051
CAS Number: 2383817-73-8