6-Chloro-2,4-difluoro-3-methylbenzoyl chloride 97%
Supplier: Apollo Scientific
PC303404-1G PC303404-250MG
APOSPC303404-1GEA 882 EUR
APOSPC303404-1G APOSPC303404-250MG
6-Chloro-2,4-difluoro-3-methylbenzoyl chloride 97%
6-Chloro-2,4-difluoro-3-methylbenzoyl chloride
Formula: C₈H₄Cl₂F₂O
MW: 225.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640058
CAS Number: 2415751-52-7