2-(2,4-Difluorophenyl)acetamide 98%
Supplier: Apollo Scientific
PC303418-1G
PC303418-25G
PC303418-5G
APOSPC303418-1GEA
142
EUR
APOSPC303418-1G
APOSPC303418-25G
APOSPC303418-5G
2-(2,4-Difluorophenyl)acetamide 98%
2-(2,4-Difluorophenyl)acetamide
Formula:
C₈H₇F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD20700103 CAS Number: 399-34-8 |