5-Bromo-2,3-difluorocinnamic acid
Supplier: Apollo Scientific
Synonyms: (E)-3-(5-bromo-2,3-difluorophenyl)acrylic acid
PC303428-1G PC303428-5G
APOSPC303428-1GEA 209 EUR
APOSPC303428-1G APOSPC303428-5G
5-Bromo-2,3-difluorocinnamic acid
5-Bromo-2,3-difluorocinnamic acid
Formula: C₉H₅BrF₂O₂
MW: 263.036 g/mol
Storage Temperature: Ambient
CAS Number: 2387460-69-5