6-Bromo-2,3-difluorobenzonitrile
Supplier: Apollo Scientific
PC303438-1G PC303438-5G
APOSPC303438-1GEA 40.6 EUR
APOSPC303438-1G APOSPC303438-5G
6-Bromo-2,3-difluorobenzonitrile
6-Bromo-2,3-difluorobenzonitrile
Formula: C₇H₂BrF₂N
MW: 217.998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381286
CAS Number: 1207875-87-3