3-Bromo-2-fluoro-5-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
PC303464-10G PC303464-1G PC303464-5G
APOSPC303464-10GEA 1040 EUR
APOSPC303464-10G APOSPC303464-1G APOSPC303464-5G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzaldehyde 98%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₃BrF₄O
MW: 271.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713159
CAS Number: 1236538-66-1