3-Bromo-2-fluoro-5-(trifluoromethyl)phenylacetonitrile
Supplier: Apollo Scientific
PC303467-1G PC303467-5G
APOSPC303467-1GEA 325 EUR
APOSPC303467-1G APOSPC303467-5G
3-Bromo-2-fluoro-5-(trifluoromethyl)phenylacetonitrile
3-Bromo-2-fluoro-5-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₄BrF₄N
MW: 282.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32265221
CAS Number: 2092565-17-6