3-Bromo-2-fluoro-5-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
PC303474-1G PC303474-5G
APOSPC303474-1GEA 410 EUR
APOSPC303474-1G APOSPC303474-5G
3-Bromo-2-fluoro-5-(trifluoromethyl)aniline 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)aniline
Formula: C₇H₄BrF₄N
MW: 258.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28740796
CAS Number: 1262198-06-0