5-Bromo-4-chloro-2-fluorobenzaldehyde 97%
Supplier: Apollo Scientific
PC303476-1G PC303476-5G
APOSPC303476-1GEA 219 EUR
APOSPC303476-1G APOSPC303476-5G
5-Bromo-4-chloro-2-fluorobenzaldehyde 97%
5-Bromo-4-chloro-2-fluorobenzaldehyde
Formula: C₇H₃BrClFO
MW: 237.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20486523
CAS Number: 1781052-25-2