5-Bromo-4-chloro-2-fluoroaniline
Supplier: Apollo Scientific
PC303485-1G PC303485-5G PC303485-250MG
APOSPC303485-1GEA 43.5 EUR
APOSPC303485-1G APOSPC303485-5G APOSPC303485-250MG
5-Bromo-4-chloro-2-fluoroaniline
5-Bromo-4-chloro-2-fluoroaniline
Formula: C₆H₄BrClFN
MW: 224.458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00274773
CAS Number: 111010-07-2