3-Bromo-5,6-difluoro-2-methoxybenzaldehyde 97%
Supplier: Apollo Scientific
PC303487-1G
PC303487-5G
PC303487-25G
PC303487-100G
APOSPC303487-1GEA
198
EUR
APOSPC303487-1G
APOSPC303487-5G
APOSPC303487-25G
APOSPC303487-100G
3-Bromo-5,6-difluoro-2-methoxybenzaldehyde 97%
3-Bromo-5,6-difluoro-2-methoxybenzaldehyde
Formula:
C₈H₅BrF₂O₂ MW: 251.025 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28629022 CAS Number: 1782614-60-1 |