3-Bromo-5,6-difluoro-2-methoxybenzoic acid
Supplier: Apollo Scientific
PC303493-1G PC303493-5G
APOSPC303493-1GEA 198 EUR
APOSPC303493-1G APOSPC303493-5G
3-Bromo-5,6-difluoro-2-methoxybenzoic acid
3-Bromo-5,6-difluoro-2-methoxybenzoic acid
Formula: C₈H₅BrF₂O₃
MW: 267.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640087
CAS Number: 1784368-01-9