3-Bromo-5,6-difluoro-2-methoxybenzonitrile
Supplier: Apollo Scientific
PC303496-1G PC303496-5G
APOSPC303496-1GEA 325 EUR
APOSPC303496-1G APOSPC303496-5G
3-Bromo-5,6-difluoro-2-methoxybenzonitrile
3-Bromo-5,6-difluoro-2-methoxybenzonitrile
Formula: C₈H₄BrF₂NO
MW: 248.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640090
CAS Number: 1891616-79-7