3-Bromo-5,6-difluoro-2-methoxyaniline
Supplier: Apollo Scientific
PC303497-1G PC303497-5G
APOSPC303497-1GEA 0 EUR
APOSPC303497-1G APOSPC303497-5G
3-Bromo-5,6-difluoro-2-methoxyaniline
3-Bromo-5,6-difluoro-2-methoxyaniline
Formula: C₇H₆BrF₂NO
MW: 238.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28653101
CAS Number: 1784147-15-4