3-Bromo-4-(trifluoromethoxy)benzylamine 98%
Supplier: Apollo Scientific
PC303527-1G PC303527-5G
APOSPC303527-1GEA 418 EUR
APOSPC303527-1G APOSPC303527-5G
3-Bromo-4-(trifluoromethoxy)benzylamine 98%
3-Bromo-4-(trifluoromethoxy)benzylamine
Formula: C₈H₇BrF₃NO
MW: 270.046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31720102
CAS Number: 1699274-98-0