3-Bromo-6-fluoro-2-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
PC303534-1G PC303534-5G
APOSPC303534-1GEA 219 EUR
APOSPC303534-1G APOSPC303534-5G
3-Bromo-6-fluoro-2-(trifluoromethyl)benzaldehyde 98%
3-Bromo-6-fluoro-2-(trifluoromethyl)benzaldehyde
Formula: C₈H₃BrF₄O
MW: 271.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640116
CAS Number: 2090471-20-6