5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetonitrile
Supplier: Apollo Scientific
PC303543-1G PC303543-5G
APOSPC303543-1GEA 337 EUR
APOSPC303543-1G APOSPC303543-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetonitrile
5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₄BrF₄N
MW: 282.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640124
CAS Number: 2092340-69-5