2-Bromo-5-fluoro-4-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
PC303552-1G PC303552-5G
APOSPC303552-1GEA 209 EUR
APOSPC303552-1G APOSPC303552-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzaldehyde 97%
2-Bromo-5-fluoro-4-(trifluoromethyl)benzaldehyde
Formula: C₈H₃BrF₄O
MW: 271.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640130
CAS Number: 2091605-30-8