2-Bromo-5-fluoro-4-(trifluoromethyl)benzyl bromide 97%
Supplier: Apollo Scientific
PC303554-1G PC303554-5G
APOSPC303554-1GEA 290 EUR
APOSPC303554-1G APOSPC303554-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzyl bromide 97%
2-Bromo-5-fluoro-4-(trifluoromethyl)benzyl bromide
Formula: C₈H₄Br₂F₄
MW: 335.919 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640132
CAS Number: 2090690-67-6